Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Montelukast Sodium Hydrate 98.0+%, TCI America™
SDP

Catalog No. M23401G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M23401G 1 g
M2340200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M23401G Supplier TCI America Supplier No. M23401G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 151767-02-1
Molecular Formula C35H35ClNNaO3S
Molecular Weight (g/mol) 608.17
MDL Number MFCD00931431
InChI Key LBFBRXGCXUHRJY-HKHDRNBDSA-M
Synonym montelukast sodium, singulair, montair, unii-u1o3j18sfl, montelukast sodium usan, montelukast na, montelukast sodium salt, u1o3j18sfl, singulair tn
PubChem CID 23663996
ChEBI CHEBI:6993
IUPAC Name sodium 2-[1-({[(1R)-1-{3-[(1E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl}-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanyl}methyl)cyclopropyl]acetate
SMILES [Na+].CC(C)(O)C1=CC=CC=C1CC[C@@H](SCC1(CC([O-])=O)CC1)C1=CC=CC(\C=C\C2=CC=C3C=CC(Cl)=CC3=N2)=C1

Specifications

Melting Point 115°C
Color White
pH 9.7 (1% aq. soln.)
Quantity 1 g
Formula Weight 608.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Montelukast Sodium Hydrate
RTECSNumber GZ0698000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.