Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moore Medical Colgate™ Softsoap™

Clinically proven to eliminate 99% of germs

Supplier:  Moore Medical 88740

Catalog No. 19-898-834

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.

This item is not returnable. View return policy

Description

Description

  • Cleans without leaving hands feeling dry
  • Dermatologist tested
  • 221.8mL (7.5 oz.)

Specifications

Specifications

Softsoap Pump Bottle
7.5 oz.
222 mL
To eliminate 99% of germs
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit