Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS 10X Solution, Fisher BioReagents™

Catalog No. BP2900500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Nalgene Poly Bottle
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP2900500 500 mL Poly Bottle
BP29001 1 L Nalgene Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP2900500

missing translation for 'validMainframeMsg'

Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA

A low ionic strength buffer used for electrophoretic separation of RNA
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2 , 7732-18-5 , 1310-73-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-n-morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-morpholinopropanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, unii-273bp63nv3, 3-n-morpholino propanesulfonic acid, morpholinopropanesulfonic acid, 3-[N-Morpholino]propane sulfonic Acid, 4-morpholinepropanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
IUPAC Name 4-(3-sulfonatopropyl)morpholin-4-ium
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Physical Form Liquid
Quantity 500 mL
DNase DNase free
Filtered Through 0.2-micron filtered
Packaging Poly Bottle
Protease Protease free
Merck Index 15, 6352
Concentration or Composition (by Analyte or Components) 10X solution contains 0.2M MOPS, 20mM sodium acetate trihydrate, 10mM disodium EDTA and NaOH to pH of 7.0
Chemical Name or Material MOPS, 10X Solution, pH 7
ShelfLife 1 year
Recommended Storage 4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.