Learn More
Description
Premixed MOPS buffer solution (pH = 7.0) at 10X concentration. A low ionic strength buffer used for electrophoretic separation of formaldehyde denatured RNA
Specifications
Specifications
| Physical Form | Liquid |
| Quantity | 500 mL |
| DNase | DNase free |
| Filtered Through | 0.2-micron filtered |
| Packaging | Poly Bottle |
| Protease | Protease free |
| Merck Index | 15, 6352 |
| Concentration or Composition (by Analyte or Components) | 10X solution contains 0.2M MOPS, 20mM sodium acetate trihydrate, 10mM disodium EDTA and NaOH to pH of 7.0 |
| Chemical Name or Material | MOPS, 10X Solution, pH 7 |
Safety and Handling
| ShelfLife | 1 year |
| Recommended Storage | 4°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.