Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, Biological Buffer, 99%, Spectrum™ Chemical
SDP

Catalog No. 18604699 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
1 kg
5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-699 50 kg Fiber Drum
18-604-695 100 g Poly Bottle
18-604-696 1 kg Poly Bottle
18-604-697 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-604-699 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. MO10250KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

MO102, 1132-61-2, C7H15NO4S

Spectrum™ Chemical MOPS, Biological Buffer

Specifications

CAS 1132-61-2
CAS Max % 1
Loss on Drying 0.01
Packaging Fiber Drum
Quantity 50 kg
Molecular Weight (g/mol) 209.26
Percent Purity 99%
Grade Reagent
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.