Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, free acid, 100.2%, MP Biomedicals™

Catalog No. ICN19483780 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19483780 100 g
ICN19483725 25 g
ICN19483790 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19483780 Supplier MP Biomedicals, Inc Supplier No. 0219483780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-[N-Morpholino]propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

For Use With (Application) Molecular Biology
Physical Form Powder
Quantity 100 g
Solution Type Buffer
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.3
Percent Purity 1.002
Grade Molecular Biology
Chemical Name or Material MOPS
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.