Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOPS, MP Biomedicals™

Catalog No. ICN10237091 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10237091 1 kg
ICN10237010 10 g
ICN10237080 100 g
ICN10237083 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10237091 Supplier MP Biomedicals, Inc Supplier No. 0210237091
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 4-morpholinepropanesulfonic acid, 3-(N-morpholino)propanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1

Specifications

Color White
For Use With (Application) Molecular Biology
Physical Form Powder
Quantity 1 kg
Solution Type Buffer
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.3
Grade Molecular Biology
Chemical Name or Material MOPS
EINECSNumber 214-478-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.