Learn More
Essendant Microfiber Cleaning Kit, 18"" Wide Blue/Green Microfiber Head, 35"" to 60"" Gray Aluminum Handle
Supplier:  Essendant BWKMFKIT
Encompass_Preferred
Boardwalk Microfiber Cleaning Kit is a complete cleaning kit that includes everything you need to start your microfiber cleaning program. Split microfibers allow for more aggressive cleaning and removal of microscopic debris than conventional cleaning methods. The compact and convenient telescopic handle extends from 35" to 60" and clips easily onto the frame. Includes colorflag coding for HACCP compliance. Cloths and pads withstand hundreds of launderings, providing durability and strength.
Catalog No. 27-911-07
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.