Learn More
Medchemexpress LLC Moracin M | 56317-21-6 | ≥98.0% | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1229425MG
Encompass_Preferred
Moracin M is a phenolic benzofuran derivative used as an analytical reference standard for research and analytical applications. It is suitable for method development, qualitative and quantitative analysis.
  • Analytical reference standard for HPLC, GC, and MS.
  • PDE4 inhibitor used in biochemical research.
  • Available in small pack sizes for assay development and method validation.
  • Soluble in common organic solvents for stock solution preparation.
Catalog No. 50-002-32412