Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA551172
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA551172 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA551172 Supplier Invitrogen™ Supplier No. PA551172
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total MORC2 protein.

Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MORC2
Gene Accession No. Q69ZX6, Q9Y6X9
Gene Alias 8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1
Gene Symbols MORC2, Morc2a
Host Species Rabbit
Immunogen Fusion protein of human MORC2.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22880, 74522
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.