Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MORF4L2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518035
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518035 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518035 Supplier Invitrogen™ Supplier No. PA518035
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with canine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

MORF4L2 is a protein coding gene. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when directly recruited to sites of DNA damage. Also component of the MSIN3A complex which acts to repress transcription by deacetylation of nucleosomal histones.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MORF4L2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene MORF4L2
Gene Accession No. Q15014
Gene Alias 2410017O14Rik; Kiaa0026; Liver regeneration-related protein LRRG00119; LRRG00119; mKIAA0026; Morf4l2; MORFL2; MORF-related gene X; MORF-related gene X protein; mortality factor 4 like 2; mortality factor 4-like protein 2; MRGX; MSL3-2 protein; OTTHUMP00000023756; protein MSL3-2; RP5-1055C14.2; Sid 393; Sid393; Sid393p; transcription factor-like protein Mrgx
Gene Symbols MORF4L2
Host Species Goat
Immunogen Synthetic peptide sequence (SSRKQGSQPRGQQS) corresponding to the N-terminus amino acids of MORF4L2 (aa 2-16).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9643
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.