Learn More
Enzo Life Sciences Morin (1g). CAS: 654055-01-3
SDP
Supplier:  Enzo Life Sciences ALX385016G001
Encompass_Preferred
Antioxidant flavonoid. Shows anti-proliferative and antitumor properties. Induces apoptosis. Induces cell cycle arrest at the G2/M phase. Anti-inflammatory compound. Suppresses NF-κB activation. Induces lipid peroxidation and DNA strand breaks. Alternative name: 2',3,4',5,7-Pentahydroxyflavone . 2H2O. Purity: ≥85% (HPLC). Appearance: Yellow to brown. Solubility: Soluble in methanol or 100% ethanol; slightly soluble in ether or acetic acid. Long Term Storage: Ambient.
Catalog No. 50-200-9046
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.