Learn More
Molecular Dimensions MORPHEUS II BUFFER SYS 4 100ML
SDP
Supplier:  Molecular Dimensions MD2100243
Encompass
Morpheus II - Buffer System 4, 100mL; Contains 100 mL bottle of untitrated 1M MOPSO and 100 mL bottle of untitrated 1M Bis-Tris.
Catalog No. NC1560471
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.