Learn More
Morphix Technologies Color Comparators for ChromAir™ Passive Sampling Systems
SDP
Use with ChromAir badge to determine the exact dose of a specific contaminant present in the workplace.
Supplier:  Morphix Technologies 384001
Encompass
Catalog No. 18-999-1386
Description
Description
Color scale identically matches the colors formed on a ChromAir badge when it is exposed to a contaminant.
  • Badge slides in comparator for comparison to color scale insert
  • Exact exposure dose is shown in the exposure dose window
Specifications
Specifications
To determine the exact dose of a specific contaminant present in the workplace
Formaldehyde
0.3 to 40 ppm.Hour
Color Comparator
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.