Learn More
eMolecules​ Morpholin-4-yl-phenyl-acetic acid | 6342-19-4 | 1MG
Supplier:  eMolecules​ 397229990
Encompass_Preferred
Matrix Scientific | Morpholin-4-yl-phenyl-acetic acid | 1MG | 6342-19-4 | MFCD09037503 | MW:218.26
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-157-3779
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.