Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Morpholine, ACS, 99.0% min

Catalog No. AA3198457
Click to view available options
Quantity:
500 g
4 x 500 g

CAS: 110-91-8 | C4H9NO | 87.12 g/mol

Morpholine is used to make other chemicals, as a corrosion inhibitor, and in detergents. It is also used as solvent and optical brightener.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydro
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5°C
Color Colorless
Density 1.007 g/mL
Boiling Point 129°C
Flash Point 35°C (95°F)
Odor Amine-like
Refractive Index 1.454
Quantity 4 x 500 g
UN Number UN2054
Beilstein 102549
Sensitivity Hygroscopic
Merck Index 14,6277
Solubility Information Soluble in water.
Formula Weight 87.12
Percent Purity ≤99.0%
Grade ACS
Physical Form Liquid
Chemical Name or Material Morpholine
Show More Show Less
Hazard Category H226-H302-H311+H331-H314-H335-H373
Hazard Statement GHS H Statement
H314-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: I; Proper Shipping Name: MORPHOLINE
EINECSNumber 203-815-1
RTECSNumber QD6475000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.