Learn More
TARGETMOL CHEMICALS INC MORRONISIDE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3414
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Morroniside would act as a regulator of hepatic inflammatory reactions and lipid metabolism in db/db mice. It exhibits protective effects against diabetic renal damage and human umbilical vein endothelial cells by inhibiting hyperglycemia and oxidative stress. Morroniside can notably protect the brain from damage induced by focal cerebral ischemia which might be related to morroniside antioxidant and anti-apoptotic properties in the brain. purity: 99%
Catalog No. 50-258-0278
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.