Description
- Low permeability
- High strength
- Hard
- High resistance to chemical attacks
- Translucent or glazed body
Specifications
Specifications
| Diameter (English) Outer | 60mm |
| Product Type | Mortar and Pestle |
| Material | Agate Stone |
| For Use With (Application) | For grinding or crushing |
| Includes | Mortar and Pestle |