Learn More
Fisherbrand™ Mortar and Pestle Deep Form
Catalog No. FB970M Shop All Fisherbrand Products
Change view
Click to view available options
Diameter (English) Outer:
60mm
70mm
80mm
90mm
100mm
110mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (English) Outer
FB970M 90mm
FB970J 60mm
FB970K 70mm
FB970L 80mm
FB970N 100mm
FB970P 110mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. FB970M
This item is not returnable. View return policy
Mortar and Pestle Deep Form
  • Low permeability
  • High strength
  • Hard
  • High resistance to chemical attacks
  • Translucent or glazed body
Specifications
Diameter (English) Outer 90mm
Product Type Mortar and Pestle Set
Material Agate Stone
For Use With (Application) For grinding or crushing
Includes Mortar and Pestle