Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MoSomatostatin Mouse Anti-Human, Alexa Fluor™ 647, Clone: U24-354, BD Pharmingen™

Catalog No. BDB570384 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570384 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB570384 Supplier BD Biosciences Supplier No. 570384
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MoSomatostatin Monoclonal antibody specifically detects MoSomatostatin in Human (QC Testing), Mouse,Rat (Lack of Reactivity Confirmed in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MoSomatostatin
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone U24-354
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias growth hormone release-inhibiting factor, SST
Host Species Mouse
Immunogen Human Preprosomatostatin Recombinant Protein
Purification Method The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6750
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.