Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOSPD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565507 Supplier Invitrogen™ Supplier No. PA565507
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLGPVVPVLVF PPDLVFRADQ RSGPRQLLTL YNPTGTALRF RVLCTAPAKY TVFDAEGYVK PQ Highest antigen sequence identity to the following orthologs - mouse 97%, rat 97%.

MOSPD3 is a multi-pass membrane protein with a major sperm protein (MSP) domain. The deletion of a similar mouse gene is associated with defective cardiac development and neonatal lethality.This gene encodes a multi-pass membrane protein with a major sperm protein (MSP) domain. The deletion of a similar mouse gene is associated with defective cardiac development and neonatal lethality. Alternate transcriptional splice variants, encoding different isoforms, have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOSPD3
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOSPD3
Gene Accession No. O75425
Gene Alias 1190005J19Rik; 5133401H10Rik; CDS3; gene trap integration gene 2; Gtig2; hypothetical protein LOC506163; Mospd3; motile sperm domain containing 3; motile sperm domain-containing protein 3; NET30; R124
Gene Symbols MOSPD3
Host Species Rabbit
Immunogen Recombinant Human MOSPD3. Recombinant protein control fragment (Product #RP-104852).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64598
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.