Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motesanib diphosphate, Thermo Scientific™

Catalog No. AC469181000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469181000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC469181000 Supplier Thermo Scientific Chemicals Supplier No. 469181000
Add to Cart
This item is not returnable. View return policy

Specifications

Chemical Name or Material Motesanib diphosphate
CAS 857876-30-3
Quantity 100 mg
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C22 H23 N5 O . 2 H3 O4 P
Physical Form Powder and/or Flakes
Color White to Light-brown
Purity ≥97.5%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.