Learn More
CPC Scientific H-Ala-Asn-Glu-Arg-Ala-Asp-Leu-Ile-Ala-Tyr-Leu-Gln-Gln-Ala-Thr-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CYTC002A
Encompass
This peptide is an antigenic peptide that prevents the development of mature 2B4 TCRhi CD4+8+ thymocytes, competitively blocks antigen induced IL-2 production plays major role in T-cell selection.ONE-LETTER SEQUENCE: ANERADLIAYLQQATKMOLECULAR FORMULA: C78H129N23O26MOLECULAR WEIGHT:1805.02STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [154277-61-9], SYNONYMS: MCC Fragment, (Gln??)-Moth Cytochrom C Fragment (88-103)RESEARCH AREA: ImmunomodulationREFERENCES: L.M.Spain et al., J. Immunol., 152, 1709 (1994)
Catalog No. 50-210-9179
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.