Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Motor Oil Composite, Restek
SDP

Catalog No. 06801766 Shop All Restek Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-801-766 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-801-766 Supplier Restek Corporation Supplier No. 31464
May include imposed supplier surcharges.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

  • Prepared from an equal-volume blend of 5W30, 10W30, 10W40, and 20W50 motor oils. After blending, a precisely weighed amount of the composite is added to a volumetric flask to produce the standard.
  • Motor oil standards may exhibit lot-to-lot variation

Specifications

Description Blend of 5W30, 10W30, 10W40, and 20W50 motor oils
Packaging Ampule
Concentration 50,000 μg/mL
Solvent or Matrix Methylene Chloride
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.