Learn More
Stoelting™ Motorized Stereotaxic Instrument for Rats and Mice
SDP
Stoelting Motorized Stereotaxic Instrument for Rats and Mice allows users to perform surgeries on both Mice and Rats on the same base
Supplier:  Stoelting™ 51770
Catalog No. 10-000-208
Only null left
Add to Cart
Request a Quote
Description
Description
  • Includes customized motors and the state-of-the-art StereoDrive software to allow motorized, computer controlled stereotaxic positioning in all 3 orthogonal axes.
  • Intuitive movement control and integrated stereotaxic atlas enables unprecedented accuracy and higher throughput in all stereotaxic applications.
Specifications
Specifications
Neuroscience
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.