Learn More
Mott Manufacturing RFV2™ Vertical Sash Fume Hood Accessory, Epoxy Worktop
SDP
Use a Mott Epoxy Worktop to protect your fume hood from damage. These come with cupsinks that on the right and designed for 18 in. deep fume hood base cabinets.
Supplier:  Mott Manufacturing RFR2660SB
 View more versions of this product
Catalog No. 01-100-197
Add to Cart
Description
Description
  • Black epoxy work surface with cupsink on right side for RFV2 Series fume hoods
  • Cupsinks are located for 18 in. deep fume hood base cabinets (not included)
Order Info
When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to.
Certifications
SEFA, ISO9001:2008
Warranty and Services
2 Years From Date of Shipment
Specifications
Specifications
RFV2™ Vertical Sash Fume Hood