Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mott Manufacturing SafeGuard™ Bench Fume Hood Accessory, Alarm
SDP

Ensure face velocity does not drop below an unsafe level. Mott SafeGuard Bench Fume Hood Alarm helps keep workers safe with clear, digital display to help monitor face velocity levels.

$1903.00

Specifications

Product Type Alarm
For Use With (Equipment) SafeGuard Bench Fume Hood
View More Specs

Products 4
Catalog Number Mfr. No. Location Price Quantity & Availability  
Catalog Number Mfr. No. Location Price Quantity & Availability  
04-000-585
View Documents
Mott Manufacturing
ALARM04SB
Each for $1,903.00
Only null left
Add to Cart
 
04-000-846
View Documents
Mott Manufacturing
ALARM04CC
Each for $1,903.00
Only null left
Add to Cart
 
04-000-237
SDP
View Documents
Mott Manufacturing
ALARM04DT
Each for $1,903.00
Only null left
Add to Cart
 
04-000-150
SDP
View Documents
Mott Manufacturing
ALARM04MGC
N/A N/A N/A
Description

Description

  • Digital display
  • Ships loose
Order Destinations
  • Zone 1 (Use CC suffix): Montana (Eastern), Minnesota, North Dakota, South Dakota, Wyoming
  • Zone 2 (Use WJ suffix): Iowa, Kansas, Missouri, Nebraska, Wisconsin
  • Zone 3 (Use SL suffix): Alabama, Florida, Georgia, Illinois, Kentucky, Tennessee
  • Zone 4 (Use NE suffix): Connecticut, Delaware, District of Columbia, Maine, Maryland, Massachusetts, New Hampshire, North Carolina, Rhode Island, South Carolina, Vermont, Virginia, West Virginia
  • Zone 5 (Previous DT suffix): Moved to Zone 9
  • Zone 6 (Previous CF suffix): Moved to Zone 9
  • Zone 7 (Use MGC suffix): Arizona, Arkansas, Colorado, Louisiana, Mississippi, New Mexico, Oklahoma, Texas
  • Zone 8 (Use S suffix): New Jersey, New York, Pennsylvania (Eastern)
  • Zone 9 (Use SV suffix): Michigan, New York State (Upper), Ohio, Pennsylvania (Western)
  • Zone 10 (Use SB suffix): Alaska, California, Hawaii, Idaho, Montana (Western), Nevada, Oregon, Utah, Washington

Order Info

When ordering, be sure to select items with the letter code suffix that matches the state that items are to be DELIVERED to

Specifications

Specifications

Alarm
SafeGuard Bench Fume Hood
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.