Learn More
United States Biological Corporation MOU IMMUNOGLOB 100TEST
Supplier:  United States Biological Corporation I750270B100TESTS
Encompass_Preferred
I7502-70B Mouse Anti-Immunoglobulin-Like Transcript 2 (ILT2, CD85j) (PE)The immunoglobulin-like transcript (ILT) family of activating and inhibitory type immunoreceptors are expressed on many leukocyte subsets and function in the regulation of immune responses (1-3). This family was also named leukocyte Ig-like receptors (LIR) and monocyte/macrophage Ig-like receptors (MIR). ILTs share significant homology with killer cell Ig-like receptors (KIR). The ILT genes are located on human chromosome 19q13.4 in the leukocyte receptor complex that also includes the genes encoding KIRs (4). With the exception of ILT6, which is a soluble molecule, all ILT family members are type I transmembrane proteins having two or four extracellular Ig-like domains (2, 3).
Catalog No. NC2308616
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.