Learn More
StatLab MOUNTING MEDIUM 1 PINT SIZE
SDP
Supplier:  StatLab MMC0143
Encompass_Preferred
Mounting Medium; Clear Mount; Master Tech; 1 pt.; Probably the fastest drying mounting medium; Clear Mount is compatible with virtually all clearing agents; Coverslipping with Clear Mount is simple, fast, and inexpensive
Catalog No. NC9202904
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.