Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Mounting Medium
SDP

Catalog No. CTS011NOV Shop All Novus Products
Change view
Click to view available options
Quantity:
15mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CTS011NOV 15mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CTS011NOV Supplier Novus Biologicals™ Supplier No. CTS011NOV

Item Discontinued Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Permanent aqueous mounting medium

  • Medium that sample is in while it is being imaged on microscope

Specifications

Content And Storage Store at Room temperature
Product Type Mounting Media
Quantity 15mL
For Use With (Application) Used as medium in microscopy

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.