Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Mounting Medium With PI, Aqueous, Anti-fade, 100 mL
Description
- Form: Liquid
- Application: ICC/IF
- Storage buffer: Preservative: 0.09% Sodium azide; Constituents: 84% Water, 0.0002% Propidium Iodide
- Storage conditions: Store at +4°C
- Shipping conditions: Blue Ice
- Description: Aqueous anti-fade mounting medium designed to preserve fluorescence when imaging tissues and cell samples
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | DNA |
| Shipping Condition | Blue Ice |
| Form | Liquid |
| Label or Dye | Propidium Iodide |
| Alternate Part Number | ab104129 |
| Buffer | 84% water, 0.0002% propidium iodide, 0.09% sodium azide |
| Content And Storage | Store at 4°C. |
| Detection Method | Fluorescence |
| Formulation | Preservative: 0.09% Sodium azide; Constituents: 84% Water, 0.0002% Propidium Iodide |
| Sample Type | Tissues, Cell samples |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?