Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Mounting Medium With PI, Aqueous, Anti-fade, 20 mL

Catalog No. 502658531
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-265-8531 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-265-8531 Supplier Abcam Supplier No. AB10412920ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Aqueous anti-fade mounting medium designed to preserve fluorescence in tissue and cell samples.

  • Applications: ICC/IF
  • Form: Liquid
  • Preservative: 0.09% Sodium azide
  • Constituents: 84% Water and 0.0002% Propidium Iodide
  • Storage Temperature: +4°C
  • Shipping Temperature: Blue Ice

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Shipping Condition Blue Ice
Format Bottle
Kit Contents Mounting Medium With PI: 20 mL
Label or Dye Propidium Iodide
Buffer 0.09% sodium azide, 84% water, 0.0002% propidium iodide
Concentration 0.0002%
Form Liquid
Detection Method Fluorescence
Alternate Part Number ab104129
Content And Storage Store at 4°C.
Formulation 0.09% sodium azide, 84% water, 0.0002% propidium iodide
Sample Type Tissues, Cell samples
Quantity 20 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.