Learn More
R&D Systems™ Mouse 20S Immunoproteasome Protein, CF

Description
Specifications
Specifications
| Host Species | Mouse |
| Purity | >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie« Blue stain |
| Content And Storage | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -70 °C as supplied. 3 months, -70 °C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Source | Mouse spleen-derived 20S Immunoproteasome protein |
| Quantity | 25 μg |
| Molecular Weight (g/mol) | 700 kDa |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.