Learn More
Description
- Also known as: 2B4; C9.1; Ly90; NAIL; Nmrk; Cd244; NKR2B4; SLAMF4; F730046O15Rik
- CD244 is a cell surface receptor expressed on natural killer (NK) cells (and some T cells) that mediate nonmajor histocompatibility complex (MHC) restricted killing.
- The interaction between NK-cell and target cells via CD244 is thought to modulate NK-cell cytolytic activity.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mCd244a |
| Accession Number | NP_061199 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1207 bp |
| Gene ID (Entrez) | 51744 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
