Learn More
American Research Products Inc Mouse a Melanocyte Stimulating
Supplier: American Research Products Inc CSBE15876M
Mouse a-Melanocyte Stimulating Hormone (a-MSH) ELISA Kit from CUSABIO Detection Range 12 5 pg/mL-800 pg/mL Sensitivity 3 12 pg/mL Sample serum plasma cell culture supernates Method Sandwich