Learn More
Biomatik Corporation Mouse Acetyl Coenzyme A Acetyltransferase 2 (ACAT2) ELISA Kit
Supplier:  Biomatik Corporation EKU02070
Encompass_Preferred
Mouse Acetyl Coenzyme A Acetyltransferase 2 (ACAT2) ELISA Kit for detecting ACAT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8CAY6. Gene ID: 110460. Target Synonyms: ACTL; Acetoacetyl Coenzyme A Thiolase; Acetyl-CoA transferase-like protein; Cytosolic acetoacetyl-CoA thiolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9156
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.