Learn More
Specifications
Specifications
| Gene | Activin RIB/ALK-4 |
| Accession Number | NP_031421 |
| Gene ID (Entrez) | 91 |
| Gene Alias | activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ActivinRIB, ActRIB, ACVR1B, ACVRLK4ACTR-IB, ALK4, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1 B |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
