Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Mouse Adiponectin Recombinant Protein, PeproTech®

Catalog No. 315261MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
250 μg
5 μg
50 μg
500 μg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
315-26-1MG 1 mg
31526100UG 100 μg
31526250UG 250 μg
315-26-25UG 25 μg
31526500UG 500 μg
315-26-50UG 50 μg
315-26-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 315-26-1MG Supplier Gibco™ Supplier No. 315261MG
Add to Cart
Add to Cart
Request a Quote
This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine Adiponectin is a multimeric glycoprotein containing amino acids Val-21 to Asn-247 of the adiponectin precursor protein fused to an N-terminal histidine tag. As a result of glycosylation, Recombinant Murine Adiponectin migrates with an apparent molecular mass of approximately 31-36 kDa by SDS-PAGE gel, under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Adiponectin is a protein hormone secreted exclusively by adipocytes that regulates the metabolism of lipids and glucose. Adiponectin has been shown to play a role in various physiological processes including homeostasis and obesity. Decreased levels of adiponectin are associated with obese individuals as well as insulin resistance and hyperinsulinemia (type 2 diabetes). In mice adiponectin has been shown to improve insulin signaling, thus ameliorating insulin resistance. Through activation of AMP kinase and PPAR-alpha, adiponectin can increase fatty acid transport, oxidation, and dissipation in skeletal muscle, leading to a reduction of intramyocellular lipid levels and thus improved insulin signaling.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q60994
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 11450
Molecular Weight (g/mol) 25.8 kDa
Name Mouse Adiponectin
Quantity 1 mg
Source Insect cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 30 kDa adipocyte complement-related protein; 30 kDa; Acdc; Acrp30; Ad; adipo; adipocyte complement related protein; adipocyte complement related protein of 30 kDa; adipocyte complement-related 30 kDa protein; adipocyte complement-related protein of 30 kDa; adipocyte, C1Q and collagen domain containing; adipocyte, C1q and collagen domain-containing protein; Adipocyte-specific protein AdipoQ; adiponectin; adiponectin, C1Q and collagen domain containing; Adipoq; Adipose most abundant gene transcript 1 protein; adipose specific collagen-like factor; ADIPQTL1; ADPIOQ; ADPN; Apm1; APM-1; APN; GBP28; Gelatin-binding protein; gelatin-binding protein 28
Common Name Adiponectin
Gene Symbol ADIPOQ
Biological Activity Determined by a cytotoxic assay using M1 cells. The ED50 for this effect is 4.0-6.0 ug/ml
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence RGHHHHHHHH VTTTEELAPA LVPPPKGTCA GWMAGIPGHP GHNGTPGRDG RDGTPGEKGE KGDAGLLGPK GETGDVGMTG AEGPRGFPGT PGRKGEPGEA AYVYRSAFSV GLETRVTVPN VPIRFTKIFY NQQNHYDGST GKFYCNIPGL YYFSYHITVY MKDVKVSLFK KDKAVLFTYD QYQEKNVDQA SGSVLLHLEV GDQVWLQVYG DGDHNGLYAD NVNDSTFTGF LLYHDTN
Content And Storage -20°C
Expression System Insect cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.