Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse aFGF ELISA Kit

Catalog No. EM5RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EM5RBX10 10 x 96 Tests
EM5RBX5 5 x 96 Tests
EM5RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EM5RBX10 Supplier Invitrogen™ Supplier No. EM5RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Mouse aFGF quantitates mouse aFGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse aFGF.

FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product
Assay Range 12 to 3,000 pg/mL
Assay Sensitivity 12 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 14164
Gene Symbol FGF1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target FGF1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.