Learn More
Description
PA1-30899 detects Mouse Albumin from mouse samples.
Specifications
Specifications
| Antigen | Mouse Albumin |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene | Alb |
| Gene Accession No. | P07724 |
| Gene Alias | Alb; Alb1; Alb-1; albumin; albumin 1; serum albumin |
| Gene Symbols | Alb |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
