Learn More
Abcam Mouse ALT ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB2828821X96TESTS
Encompass_Preferred
Mouse ALT ELISA Kit is a single-wash, 90-minute SimpleStep ELISA used to quantify mouse alanine aminotransferase (ALT) with a sensitivity of 4 pg/mL. The assay follows a simple Mix-Wash-Read protocol with just one incubation and wash step. - Colorimetric Sandwich ELISA – 450 nm readout, compatible with any standard plate reader - Different formats for different needs – 384-well format available for high-throughput applications - Broad sample compatibility – works with serum, plasma - Cited in over 15 publications
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2015921