Learn More
ACROBiosystems ACROBiosystems Mouse Angiopoietin-like 3 / ANGPTL3 Protein, His Tag
Supplier:  ACROBiosystems AN3M52H31MG
Encompass_Preferred
Recombinant Protein;1MG;Mouse Angiopoietin-like 3, His Tag (AN3-M52H3) is expressed from human 293 cells (HEK293). It contains AA Ser 17 - Thr 455 (Accession # Q9R182-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 52.6 kDa.
Catalog No. 50-210-6905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.