Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Mouse ANGPTL2 Fc Chimera Recombinant Protein

Catalog No. PIRP88201
Change view
Click to view available options
Quantity:
50 μg
500 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88201 50 μg
PIRP88202 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PIRP88201 Supplier Invitrogen™ Supplier No. RP88201
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (705 μL) be added to the vial to prepare a stock solution of 0.71 mg/mL. Concentration is measured by UV.

Angiopoietins are members of the vascular endothelial growth factor family and the only known growth factors largely specific for vascular endothelium. Angiopoietin-1, angiopoietin-2, and angiopoietin-4 participate in the formation of blood vessels. The protein encoded by this gene is another member of the angiopoietin family that is widely expressed in adult tissues with mRNA levels highest in highly vascularized tissues. This protein was found to be a secretory protein that does not act as an endothelial cell mitogen in vitro.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9R045
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 26360
Molecular Weight (g/mol) 58 kDa
Name Mouse ANGPTL2 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias AI593246; angiopoietin like 2; angiopoietin related protein 2; angiopoietin-like 2; angiopoietin-like protein 2; Angiopoietin-related protein 2; angiopoietin-related protein-2; Angptl2; ARP2; AW260363; HARP; MGC8889; UNQ170/PRO196
Common Name ANGPTL2
Gene Symbol ANGPTL2
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Mouse ANGPTL2, amino acids Asp245-His493 (Accession # Q9R045) with an N-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.