Learn More
Description
- Angptl8 angiopoietin-like 8 [ Mus musculus (house mouse) ]
- Also known as: Rifl; Gm6484; EG624219
- ANGPTL8/Betatrophin is a secreted hormone protein
- It is primarily expressed in the liver and adipose tissue and acts as a blood lipid regulator by regulating serum triglyceride levels
- Betatrophin has also been shown to be increased in the plasma of diabetic patients
- The Angiopoietin-like protein 8/Betatrophin insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mAngptl8 |
| Accession Number | NP_001074409 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 610 bp |
| Gene ID (Entrez) | 55908 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
