Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ADNP, Mouse, Clone: 6B8, Abnova™

Catalog No. 89106596 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-596 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-596 Supplier Abnova Corporation Supplier No. H00023394M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ADNP.

Vasoactive intestinal peptide is a neuroprotective factor that has a stimulatory effect on the growth of some tumor cells and an inhibitory effect on others. This gene encodes a protein that is upregulated by vasoactive intestinal peptide and may be involved in its stimulatory effect on certain tumor cells. The encoded protein contains one homeobox and nine zinc finger domains, suggesting that it functions as a transcription factor. This gene is also upregulated in normal proliferative tissues. Finally, the encoded protein may increase the viability of certain cell types through modulation of p53 activity. Alternatively spliced transcript variants encoding the same protein have been described. [provided by RefSeq

Sequence: TMQGDREQLKWKNSSYGKVEGFWSKDQSQWKNASENDERLSNPQIEWQNSTIDSEDGEQFDNMTDGVAEPMHGSLAGVKLSSQQA

Specifications

Antigen ADNP
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ADNP.
Formulation ascites with no preservative
Gene ADNP
Gene Accession No. NM_015339
Gene Alias ADNP1/KIAA0784
Gene Symbols ADNP
Host Species Mouse
Immunogen ADNP (NP_056154,1018 a.a. ∽ 1102 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23394
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.