Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AFAP1, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89107238 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-238 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-238 Supplier Abnova Corporation Supplier No. H00060312B01P
Only null left
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human AFAP1 protein.

The protein encoded by this gene is a Src binding partner. It may represent a potential modulator of actin filament integrity in response to cellular signals, and may function as an adaptor protein by linking Src family members and/or other signaling proteins to actin filaments. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MEELIVELRLFLELLDHEYLTSTVREKKAVITNILLRIQSSKGFDVKDHAQKQETANSLPAPPQMPLPEIPQPWLPPDSGPPPLPTSSLPEGYYEEAVPLSPGKAPEYITSNYDSDAMSSSYESYDEEEEDGKGKKTRHQWPSEEASMDLVKDAKICAFLLRKKRFGQWTKLLCVIKDTKLLCYKSSKDQQPQMELPLQGCNITYIPKDSKKKKHELKITQQGTDPLVLAVQSKEQAEQWLKVIKEAYSGCSGPVDSECPPPPSSPVHKAELEKKLSSERPSSDGEGVVENGITTCNGKEQVKRKKSSKSEAKGTVSKVTGKKITKIISLGKKKPSTDEQTSSAEEDVPTCGYLNVLSNSRWRERWCRVKDNKLIFHKDRTDLKTHIVSIPLRGCEVIPGLDCKHPLTFRLLRNGQEVAVLEASSSEDMGRWIGILLAETGSSTDPEALHYDYIDVEMSASVIQTAKQTFCFMNRRVISANPYLGGTSNGYAHPSGTALHYDDVPCINGSLKGKKPPVASNGVTGKGKTLSSQPKKADPAAVVKRTGSNAAQYKYGKNRVEADAKRLQTKEEELLKRKEALRNRLAQLRKERKDLRAAIEVNAGRKPQAILEEKLKQLEEECRQKEAERVSLELELTEVKESLKKALAGGVTLGLAIEPKSGTSSPQSPVFRHRTLENSPISSCDTSDTEGPVPVNSAAVLKKSQAAPGSSPCRGHVLRKAKEWELKNGT

Specifications

Antigen AFAP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human AFAP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene AFAP1
Gene Accession No. BC032777.1
Gene Alias AFAP/AFAP-110/FLJ56849
Gene Symbols AFAP1
Host Species Mouse
Immunogen AFAP1 (AAH32777.1,1 a.a. ∽ 730 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60312
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.