Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHMT2, Mouse, Clone: 1E8, Abnova™

Catalog No. 89106644 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-644 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-644 Supplier Abnova Corporation Supplier No. H00023743M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BHMT2.

Homocysteine is a sulfur-containing amino acid that plays a crucial role in methylation reactions. Transfer of the methyl group from betaine to homocysteine creates methionine, which donates the methyl group to methylate DNA, proteins, lipids, and other intracellular metabolites. The protein encoded by this gene is one of two methyl transferases that can catalyze the transfer of the methyl group from betaine to homocysteine. Anomalies in homocysteine metabolism have been implicated in disorders ranging from vascular disease to neural tube birth defects such as spina bifida. [provided by RefSeq

Sequence: MAPAGRPGAKKGILERLESGEVVIGDGSFLITLEKRGYVKAGLWTPEAVIEHPDAVRQLHMEFLRAGSNVMQTFTFSASEDNMESKWEDVNAAACDLAREVAGKGDALVA

Specifications

Antigen BHMT2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BHMT2.
Formulation ascites with no preservative
Gene BHMT2
Gene Accession No. NM_017614
Gene Alias FLJ20001
Gene Symbols BHMT2
Host Species Mouse
Immunogen BHMT2 (NP_060084,1 a.a. ∽ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23743
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.