Learn More
Mouse anti-Bovine IgA Heavy Chain Secondary Antibody (IL-A71), DyLight 650, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgA Heavy Chain |
| Applications | ELISA, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IL-A71 |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Host Species | Mouse |
| Immunogen | Purified bovine Immunoglobulin |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.