Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Bovine IgG (H+L) Secondary Antibody (IL-A2), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB554853 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB554853 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB554853 Supplier Novus Biologicals Supplier No. NBP268413V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IgG (H+L) Monoclonal specifically detects IgG (H+L) in Bovine samples. It is validated for ELISA, Immunoprecipitation.

Specifications

Antigen IgG (H+L)
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone IL-A2
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Purified bovine IgG
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.