Learn More
Mouse anti-Bovine IgG2 Heavy Chain Secondary Antibody (IL-A73), FITC, R&D Systems™
Shop All Novus Products

Description
Specifications
Specifications
| Antigen | IgG2 Heavy Chain |
| Applications | ELISA, Flow Cytometry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | IL-A73 |
| Conjugate | FITC |
| Formulation | PBS |
| Host Species | Mouse |
| Immunogen | Purified bovine IgG2 |
| Purification Method | Protein A purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.